Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-2-chloro-D-phenylalanine 97%

Supplier: AMBEED, INC
Warning

A156177-1G A156177-250MG A156177-25G A156177-5G
76871-864EA 16 USD InStock
76871-864 76871-866 76871-868 76871-870
Boc-2-chloro-D-phenylalanine 97%
Boc-2-chloro-D-phenylalanine
Precaución: for Research Use Only


Formula: C₁₄H₁₈ClNO₄
MDL Number: MFCD00801056
CAS Number: 80102-23-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty