Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-3,4-diphenylisoxazole 97%

Supplier: AMBEED, INC
Warning

A156572-100G A156572-10G A156572-1G A156572-25G A156572-5G
76871-882EA 127 USD InStock
76871-882 76871-884 76871-886 76871-888 76871-890
5-Methyl-3,4-diphenylisoxazole 97%
5-Methyl-3,4-diphenylisoxazole
Precaución: for Research Use Only


Formula: C₁₆H₁₃NO
MW: 235.29 g/mol
MDL Number: MFCD18449672
CAS Number: 37928-17-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty