Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methylpyridine 1-oxide 95%

Supplier: AMBEED, INC
Warning

A157008-1G A157008-250MG A157008-5G
76871-894EA 89 USD InStock
76871-894 76871-896 76871-898
2-Chloro-5-methylpyridine 1-oxide 95%
2-Chloro-5-methylpyridine 1-oxide
Precaución: for Research Use Only


Formula: C₆H₆ClNO
MW: 143.571 g/mol
MDL Number: MFCD11977414
CAS Number: 20173-49-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty