Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(Diethylamino)-4-methylcoumarin 98%

Supplier: AMBEED, INC
Warning

A157907-1000G A157907-100G A157907-10G A157907-25G
76871-920EA 198 USD InStock
76871-920 76871-922 76871-924 76871-926 76871-928
7-(Diethylamino)-4-methylcoumarin 98%
7-(Diethylamino)-4-methylcoumarin
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₂
MDL Number: MFCD00006864
CAS Number: 91-44-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty