Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Phenoxypropionic acid 98%

Supplier: AMBEED, INC
Warning

A160032-100G A160032-10G A160032-5G
76871-972EA 113 USD InStock
76871-972 76871-974 76871-976
(±)-2-Phenoxypropionic acid 98%
(±)-2-Phenoxypropionic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00002643
CAS Number: 940-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty