Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chloropyridine 97%

Supplier: AMBEED, INC
Danger

A163916-100G A163916-10G A163916-1G A163916-5G A163916-25G
76872-112EA 139 USD InStock
76872-112 76872-114 76872-116 76872-118 77611-596
3-Bromo-5-chloropyridine 97%
3-Bromo-5-chloropyridine
Precaución: for Research Use Only


Formula: C₅H₃BrClN
MDL Number: MFCD04114221
CAS Number: 73583-39-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty