Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2H-indazole-4-boronic acid 98%

Supplier: AMBEED, INC
Warning

A168600-100MG A168600-1G A168600-250MG
76872-304EA 45 USD InStock
76872-304 76872-306 76872-308
2-Methyl-2H-indazole-4-boronic acid 98%
2-Methyl-2H-indazole-4-boronic acid
Precaución: for Research Use Only


Formula: C₈H₉BN₂O₂
MDL Number: MFCD09870055
CAS Number: 1001907-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty