Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Fluoro-2-quinolinol 97%

Supplier: AMBEED, INC
Warning

A168988-100MG A168988-1G A168988-250MG A168988-5G
76872-320EA 16 USD InStock
76872-320 76872-322 76872-324 76872-326
8-Fluoro-2-quinolinol 97%
8-Fluoro-2-quinolinol
Precaución: for Research Use Only


Formula: C₉H₆FNO
MDL Number: MFCD12026113
CAS Number: 71738-83-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty