Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetonitrile 97%

Supplier: AMBEED, INC
Warning

A172145-10G A172145-1G A172145-5G A172145-25G
76872-458EA 27 USD InStock
76872-458 76872-460 76872-462 77671-448
2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetonitrile 97%
2-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₉H₅F₂NO₂
MW: 197.13 g/mol
MDL Number: MFCD09923942
CAS Number: 68119-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty