Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Methyl-5-thiazolyl)ethyl butyrate 95%

Supplier: AMBEED, INC
Warning

A172201-100G A172201-25G A172201-5G
76872-464EA 126 USD InStock
76872-464 76872-466 76872-468
2-(4-Methyl-5-thiazolyl)ethyl butyrate 95%
2-(4-Methyl-5-thiazolyl)ethyl butyrate
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂S
MDL Number: MFCD09032913
CAS Number: 94159-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty