Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ixazomib 98%

Supplier: AMBEED, INC
Warning

A172813-250MG A172813-5MG
76872-478EA 424 USD InStock
76872-478 76872-480
Ixazomib 98%
Ixazomib
Precaución: for Research Use Only


Formula: C₁₄H₁₉BCl₂N₂O₄
MDL Number: MFCD18251438
CAS Number: 1072833-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty