Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-6-methoxypyridine 97%

Supplier: AMBEED, INC
Warning

A178074-100MG A178074-1G A178074-250MG
76872-638EA 28 USD InStock
76872-638 76872-640 76872-642
2,4-Dichloro-6-methoxypyridine 97%
2,4-Dichloro-6-methoxypyridine
Precaución: for Research Use Only


Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
MDL Number: MFCD16610793
CAS Number: 1227572-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty