Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1,2-difluoro-3-nitrobenzene 95%

Supplier: AMBEED, INC
Warning

A178434-10G A178434-1G A178434-250MG A178434-5G
76872-646EA 65 USD InStock
76872-646 76872-648 76872-650 76872-652
5-Bromo-1,2-difluoro-3-nitrobenzene 95%
5-Bromo-1,2-difluoro-3-nitrobenzene
Precaución: For research use only.


Formula: C₆H₂BrF₂NO₂
MW: 237.986 g/mol
MDL Number: MFCD18311821
CAS Number: 1261988-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty