Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2,3-Dihydro-1H-inden-1-amine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A182147-10G A182147-1G A182147-25G A182147-5G
76872-782EA 40 USD InStock
76872-782 76872-784 76872-786 76872-788
(S)-2,3-Dihydro-1H-inden-1-amine hydrochloride 97%
(S)-2,3-Dihydro-1H-inden-1-amine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClN
MDL Number: MFCD16661198
CAS Number: 32457-23-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty