Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichlorobenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A186349-1000G A186349-100G A186349-10G A186349-500G A186349-25G
76872-908EA 67 USD InStock
76872-908 76872-910 76872-912 76872-914 77084-166
2,6-Dichlorobenzaldehyde 97%
2,6-Dichlorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄Cl₂O
MDL Number: MFCD00003307
CAS Number: 83-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty