Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-α-Aminoadipic acid 98%

Supplier: AMBEED, INC
Warning

A187348-100G A187348-1G A187348-25G A187348-5G
76872-954EA 155 USD InStock
76872-954 76872-956 76872-958 76872-960
D-α-Aminoadipic acid 98%
D-α-Aminoadipic acid
Precaución: for Research Use Only


Formula: C₆H₁₁NO₄
MDL Number: MFCD00063118
CAS Number: 7620-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty