Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chloro-3-fluoropyridine 98%

Supplier: AMBEED, INC
Warning

A188596-100G A188596-10G A188596-1G A188596-250MG A188596-25G A188596-5G
76872-990EA 195 USD InStock
76872-990 76872-992 76872-994 76872-996 76872-998 76873-000
5-Bromo-2-chloro-3-fluoropyridine 98%
5-Bromo-2-chloro-3-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrClFN
MW: 210.43
MDL Number: MFCD03411574
CAS Number: 831203-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty