Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,4-Dimethoxyphenyl)acetonitrile 98%

Supplier: AMBEED, INC
Warning

A193716-100G A193716-10G A193716-25G
76873-132EA 26 USD InStock
76873-132 76873-134 76873-136
(3,4-Dimethoxyphenyl)acetonitrile 98%
(3,4-Dimethoxyphenyl)acetonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00001911
CAS Number: 93-17-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty