Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,5-dihydrothiazole-4-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A194716-100G A194716-10G A194716-25G A194716-5G
76873-184EA 103 USD InStock
76873-184 76873-186 76873-188 76873-190 76873-192
2-Amino-4,5-dihydrothiazole-4-carboxylic acid 97%
2-Amino-4,5-dihydrothiazole-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₄H₆N₂O₂S
MDL Number: MFCD00010067
CAS Number: 2150-55-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty