Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butyl 4-ethyl 4-(aminomethyl)piperidine-1,4-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A195083-100MG A195083-1G A195083-250MG A195083-5G
76873-216EA 19 USD InStock
76873-216 76873-218 76873-220 76873-222
1-tert-Butyl 4-ethyl 4-(aminomethyl)piperidine-1,4-dicarboxylate 97%
1-tert-Butyl 4-ethyl 4-(aminomethyl)piperidine-1,4-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₄H₂₆N₂O₄
MW: 286.37 g/mol
MDL Number: MFCD14525454
CAS Number: 1016258-69-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty