Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Imidazol-4-yl)methanol hydrochloride 97%

Supplier: AMBEED, INC
Warning

A200971-100G A200971-10G A200971-1G A200971-5G
76873-386EA 191 USD InStock
76873-386 76873-388 76873-390 76873-392
(1H-Imidazol-4-yl)methanol hydrochloride 97%
(1H-Imidazol-4-yl)methanol hydrochloride
Precaución: for Research Use Only


Formula: C₄H₇ClN₂O
MDL Number: MFCD00012697
CAS Number: 32673-41-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty