Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Arginine L-aspartate 97%

Supplier: AMBEED, INC
Warning

A201132-1000G A201132-100G A201132-10G A201132-25G A201132-500G
76873-398EA 81 USD InStock
76873-398 76873-400 76873-402 76873-404 76873-406
L-Arginine L-aspartate 97%
L-Arginine L-aspartate
Precaución: for Research Use Only


Formula: C₁₀H₂₁N₅O₆
MDL Number: MFCD00058334
CAS Number: 7675-83-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty