Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Leucine ethyl ester hydrochloride 95%

Supplier: AMBEED, INC
Warning

A203678-100G A203678-25G A203678-5G
76873-478EA 154 USD InStock
76873-478 76873-480 76873-482
D-Leucine ethyl ester hydrochloride 95%
D-Leucine ethyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₇NO₂·HCl
MDL Number: MFCD00190936
CAS Number: 73913-65-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty