Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-4-pyridineboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A204328-1G A204328-250MG A204328-25G A204328-5G
76873-520EA 17 USD InStock
76873-520 76873-522 76873-524 76873-526
2,6-Dichloro-4-pyridineboronic acid pinacol ester 98%
2,6-Dichloro-4-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₄BCl₂NO₂
MDL Number: MFCD09746195
CAS Number: 408492-27-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty