Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%

Supplier: AMBEED, INC
Warning

A204884-1G A204884-250MG A204884-25G A204884-5G
76873-534EA 7 USD InStock
76873-534 76873-536 76873-538 76873-540
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₂ClF₃IN
MW: 307.44 g/mol
MDL Number: MFCD10699394
CAS Number: 205444-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty