Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Triisopropylphenylboronic acid dimethyl ester 95+%

Supplier: AMBEED, INC
Warning

A205630-1G A205630-250MG A205630-5G
76873-566EA 207 USD InStock
76873-566 76873-568 76873-570
2,4,6-Triisopropylphenylboronic acid dimethyl ester 95+%
2,4,6-Triisopropylphenylboronic acid dimethyl ester
Precaución: for Research Use Only


Formula: C₁₇H₂₉BO₂
MW: 276.2220 g/mol
MDL Number: MFCD09265170
CAS Number: 145434-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty