Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Tryptophan ethyl ester hydrochloride 97%

Supplier: AMBEED, INC
Warning

A213156-100G A213156-10G A213156-1G A213156-5G
76873-732EA 55 USD InStock
76873-732 76873-734 76873-736 76873-738
D-Tryptophan ethyl ester hydrochloride 97%
D-Tryptophan ethyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₆N₂O₂·HCl
MW: 268.741 g/mol
MDL Number: MFCD00067552
CAS Number: 61535-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty