Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dulcoside B 98%

Supplier: AMBEED, INC
Warning

A219420-100MG A219420-10MG A219420-1G A219420-1MG A219420-250MG A219420-50MG A219420-5G A219420-5MG
76873-882EA 46 USD InStock
76873-882 76873-884 76873-886 76873-888 76873-890 76873-892 76873-894 76873-896
Dulcoside B 98%
Dulcoside B
Precaución: for Research Use Only


Formula: C₄₄H₇₀O₂₂
MDL Number: MFCD02752304
CAS Number: 63550-99-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty