Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-iodo-4,5-dimethoxybenzoate 97%

Supplier: AMBEED, INC
Warning

A221768-100MG A221768-1G A221768-250MG A221768-5G
76873-970EA 6 USD InStock
76873-970 76873-972 76873-974 76873-976
Methyl-2-iodo-4,5-dimethoxybenzoate 97%
Methyl 2-iodo-4,5-dimethoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁IO₄
MDL Number: MFCD09800786
CAS Number: 173043-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty