Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-dichloro-3-methylaniline 98%

Supplier: AMBEED, INC
Danger

A222601-1G A222601-250MG A222601-5G
76874-002EA 36 USD InStock
76874-002 76874-004 76874-006
4-Bromo-2,6-dichloro-3-methylaniline 98%
4-Bromo-2,6-dichloro-3-methylaniline
Precaución: for Research Use Only


Formula: C₇H₆BrCl₂N
MW: 254.9400 g/mol
MDL Number: MFCD09800852
CAS Number: 62406-68-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty