Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

I-BET762 98+%

Supplier: AMBEED, INC
Warning

A222643-10MG A222643-1MG A222643-2MG A222643-50MG A222643-5MG
76874-008EA 40 USD InStock
76874-008 76874-010 76874-012 76874-014 76874-016
I-BET762 98+%
I-BET762
Precaución: for Research Use Only


Formula: C₂₂H₂₂ClN₅O₂
MDL Number: MFCD22417091
CAS Number: 1260907-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty