Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

((S)-Piperidin-2-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A223568-100G A223568-10G A223568-1G A223568-25G A223568-5G
76874-032EA 759 USD InStock
76874-032 76874-034 76874-036 76874-038 76874-040
((S)-Piperidin-2-yl)methanol 97%
((S)-Piperidin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₆H₁₃NO
MDL Number: MFCD11036291
CAS Number: 41373-39-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty