Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-8055 98%

Supplier: AMBEED, INC
Warning

A224432-10MG A224432-1MG A224432-100MG A224432-250MG
76874-056EA 37 USD InStock
76874-056 76874-058 77671-982 77671-984
AZD-8055 98%
AZD-8055
Precaución: for Research Use Only


Formula: C₂₅H₃₁N₅O₄
MW: 465.5 g/mol
MDL Number: MFCD16660191
CAS Number: 1009298-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty