Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-nitronaphthalene 96%

Supplier: AMBEED, INC
Warning

A225228-100MG A225228-1G A225228-5G A225228-250MG
76874-066EA 9 USD InStock
76874-066 76874-068 76874-070 77671-988
1-Bromo-4-nitronaphthalene 96%
1-Bromo-4-nitronaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₆BrNO₂
MDL Number: MFCD00971828
CAS Number: 4236-05-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty