Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1-methylimidazole 96%

Supplier: AMBEED, INC
Warning

A226452-100MG A226452-1G A226452-250MG A226452-5G
76874-130EA 12 USD InStock
76874-130 76874-132 76874-134 76874-136
2-Chloro-1-methylimidazole 96%
2-Chloro-1-methylimidazole
Precaución: for Research Use Only


Formula: C₄H₅ClN₂
MDL Number: MFCD02179531
CAS Number: 253453-91-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty