Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(1-Methylpiperidin-2-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A226507-100MG A226507-1G A226507-250MG A226507-5G
76874-138EA 8 USD InStock
76874-138 76874-140 76874-142 76874-144
(S)-(1-Methylpiperidin-2-yl)methanol 95%
(S)-(1-Methylpiperidin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD20489103
CAS Number: 136030-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty