Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfachloropyridazine 95%

Supplier: AMBEED, INC
Warning

A227322-100MG A227322-1G A227322-250MG A227322-25G A227322-5G
76874-164EA 5 USD InStock
76874-164 76874-166 76874-168 76874-170 77672-002
Sulfachloropyridazine 95%
Sulphachloropyridazine
Precaución: for Research Use Only


Formula: C₁₀H₉ClN₄O₂S
MDL Number: MFCD00057371
CAS Number: 80-32-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty