Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-cyanobenzoic acid 97%

Supplier: AMBEED, INC
Danger

A227438-1G A227438-250MG
76874-172EA 35 USD InStock
76874-172 76874-174
4-Bromo-3-cyanobenzoic acid 97%
4-Bromo-3-cyanobenzoic acid
Precaución: for Research Use Only


Formula: C₈H₄BrNO₂
MW: 226.02 g/mol
MDL Number: MFCD11977390
CAS Number: 887757-25-7
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty