Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Acetamido-3-chloropropan-2-yl acetate 97%

Supplier: AMBEED, INC
Warning

A228932-100G A228932-1G A228932-25G
76874-200EA 57 USD InStock
76874-200 76874-202 76874-204 76874-206
(S)-1-Acetamido-3-chloropropan-2-yl acetate 97%
(S)-1-Acetamido-3-chloropropan-2-yl acetate
Precaución: for Research Use Only


Formula: C₇H₁₂ClNO₃
MDL Number: MFCD09842277
CAS Number: 183905-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty