Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dibenzylamino)cyclohexanone hydrochloride 95%

Supplier: AMBEED, INC
Warning

A228954-1G A228954-250MG
76874-208EA 36.08 USD InStock
76874-208 76874-210 76874-212
4-(Dibenzylamino)cyclohexanone hydrochloride 95%
4-(Dibenzylamino)cyclohexanone hydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₂₄ClNO
MW: 329.86 g/mol
MDL Number: MFCD22421636
CAS Number: 1416352-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty