Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiodiglycolic anhydride 97%

Supplier: AMBEED, INC
Warning

A236342-100G A236342-10G A236342-25G
76874-326EA 23 USD InStock
76874-326 76874-328 77304-042
Thiodiglycolic anhydride 97%
Thiodiglycolic anhydride
Precaución: for Research Use Only


Formula: C₄H₄O₃S
MW: 132.13 g/mol
MDL Number: MFCD00051689
CAS Number: 3261-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty