Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1H-imidazo[4,5-b]pyridine-6-carboxylate 98%

Supplier: AMBEED, INC
Warning

A242751-100MG A242751-250MG
76874-442EA 10 USD InStock
76874-442 76874-444
Methyl 1H-imidazo[4,5-b]pyridine-6-carboxylate 98%
Methyl 1H-imidazo[4,5-b]pyridine-6-carboxylate
Precaución: for Research Use Only


Formula: C₈H₇N₃O₂
MDL Number: MFCD13176754
CAS Number: 77862-95-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty