Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyano-5-(trifluoromethyl)benzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A243901-100MG A243901-1G A243901-250MG
76874-472EA 37 USD InStock
76874-472 76874-474 76874-476
3-Cyano-5-(trifluoromethyl)benzeneboronic acid 98%
3-Cyano-5-(trifluoromethyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₈H₅BF₃NO₂
MW: 214.938 g/mol
MDL Number: MFCD03095102
CAS Number: 1212021-62-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty