Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,2-difluoropent-4-enoate 98%

Supplier: AMBEED, INC
Danger

A244128-1G A244128-25G A244128-5G
76874-486EA 19 USD InStock
76874-486 76874-488 77672-186
Ethyl 2,2-difluoropent-4-enoate 98%
Ethyl 2,2-difluoropent-4-enoate
Precaución: for Research Use Only


Formula: C₇H₁₀F₂O₂
MDL Number: MFCD09763639
CAS Number: 110482-96-7
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty