Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl-3-fluoro-4-oxopiperidine-1-carboxylate 96%

Supplier: AMBEED, INC
Warning

A252863-10G A252863-1G A252863-250MG A252863-25G A252863-5G
76874-654EA 179 USD InStock
76874-654 76874-656 76874-658 76874-660 76874-662
Benzyl-3-fluoro-4-oxopiperidine-1-carboxylate 96%
Benzyl-3-fluoro-4-oxopiperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₄FNO₃
MDL Number: MFCD11976639
CAS Number: 845256-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty