Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-4-methoxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A254463-100MG A254463-10G A254463-1G A254463-250MG
76874-686EA 9 USD InStock
76874-686 76874-688 76874-690 76874-692
2-Amino-5-bromo-4-methoxybenzoic acid 95%
2-Amino-5-bromo-4-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₈BrNO₃
MDL Number: MFCD21868786
CAS Number: 169045-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty