Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate 97%

Supplier: AMBEED, INC
Warning

A263244-100G A263244-10G A263244-500G
76874-930EA 25 USD InStock
76874-930 76874-932 76874-934
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate 97%
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Precaución: for Research Use Only


Formula: C13H13N3O4
MW: 275.2600 g/mol
MDL Number: MFCD02947731
CAS Number: 89392-03-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty