Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1H-pyrazolo[4,3-d]pyrimidine 95%

Supplier: AMBEED, INC
Warning

A274089-100MG A274089-1G A274089-250MG A274089-5G
76875-154EA 13 USD InStock
76875-154 76875-156 76875-158 76875-160
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine 95%
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Precaución: for Research Use Only


Formula: C₅H₃ClN₄
MDL Number: MFCD18251003
CAS Number: 633328-98-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty