Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrolidine-3-carbonitrile hydrochloride 97%

Supplier: AMBEED, INC
Warning

A274321-100MG A274321-1G A274321-250MG
76875-162EA 230 USD InStock
76875-162 76875-164 76875-166
Pyrrolidine-3-carbonitrile hydrochloride 97%
Pyrrolidine-3-carbonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉ClN₂
MW: 132.59 g/mol
MDL Number: MFCD09027325
CAS Number: 1187930-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty