Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-hydroxybenzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A276663-100MG A276663-1G A276663-250MG A276663-25G A276663-5G
76875-198EA 5 USD InStock
76875-198 76875-200 76875-202 76875-204 76875-206
2-Chloro-6-hydroxybenzeneboronic acid 98%
2-Chloro-6-hydroxybenzeneboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BClO₃
MDL Number: MFCD18393225
CAS Number: 958646-70-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty